Would you like to log in again?
Nunc® Lab-Tek™ II Chambered Coverglass, Thermo Scientific
Supplier: Thermo Scientific
155409 155409 155379 155360 155379 155360 155382 155382
62407-056CS 1663.17 USD InStock
62407-056 CA62407-054 62407-054 CA62407-056 CA-62407-052 62407-052 CA62407-050 62407-050
Nunc® Lab-Tek™ II Chambered Coverglass, Thermo Scientific
Cover Glasses
Culture and view living cells on a single device
Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass includes a thin coverglass that provides the optimum optical characteristics required for high-magnification microscopy and confocal image analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty