Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nunc® Lab-Tek™ Chamber Slide™ Systems, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nunc® Lab-Tek™
177429 177402 177437 177402 177429 177445 177399 177380 178599 177410 177380 177410 177399 177437 177445 178599
62407-320CS 1242.1 USD InStock
62407-320 62407-330 CA62407-350 62407-350 62407-335 CA62407-310 CA62407-320 CA62407-331 62407-310 CA62407-335A CA62407-325 62407-305 62407-315 62407-325 CA62407-317 CA62407-305
Nunc® Lab-Tek™ Chamber Slide™ Systems, Sterile, Thermo Scientific
Slides Chamber Slides
Systems feature a removable polystyrene medium chamber so cells can be grown, fixed, and stained on the same slide.
  • Removable media wells are attached to either a soda-lime glass microscope slide or a solvent-resistant Permanox® plastic slide by a medical-grade silicone gasket
  • A convenient writing surface is easily marked, providing positive sample identification
  • Multiwell design allows researchers to analyze different specimens, immunochemicals, or stains on one slide without transfer from other vessels
Tested for quality and sterility.
Packaging: Packed sterile, 8 slides per incubator tray.
Caution: Do not use mounting media containing toluene or xylene on Permanox slides.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty