Would you like to log in again?
Nunc® MicroWell™ 60- and 72-Well Mini Trays, Thermo Scientific
Supplier: Thermo Scientific
Nunc® MicroWekk™
470378 438733 472400 452256 438733 472400 470378 439225 452256 439225
62407-126CS 1571.58 USD InStock
CA62407-382 62407-126 CA62409-297 CA62407-127 CA62407-126 CA62409-296 62407-127 62409-297 62409-296 62407-382
Nunc® MicroWell™ 60- and 72-Well Mini Trays, Thermo Scientific
Microplates Immunology Microplates and Strips
These Terasaki-style mini trays are ideal for serotyping, HLA applications, microlymphocytotoxicity, cell cloning studies, protein crystallization, and sequencing reactions.
Tray wells have a special polystyrene surface treatment to ensure enhanced mixing of reagents and samples in each well, even when frozen and thawed. Treatment ensures a limited affinity to antibodies even in high concentrations and limited protein adsorption. The optical clarity of the trays allows them to be viewed with an inverted microscope. Trays feature large raised letters for easy identification. Wells feature an inverted, truncated cone shape and a working volume of 10µL.
High- and low-profile trays are stackable when included lid is used. S-plates are stackable without a lid.
Ordering information: High- and low-profile trays are supplied with a uni-directional lid.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty