Would you like to log in again?
Nunc® Disposable Tubes, Round-Bottom, Polystyrene, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
342919 343141 343923 343141 342919 343923
62409-459CS 500.9 USD InStock
CA62409-459 62409-459 CA62409-467 CA62409-465 62409-467 62409-465
Nunc® Disposable Tubes, Round-Bottom, Polystyrene, Thermo Scientific
Tubes Centrifuge Tubes
These transparent, polystyrene tubes offer a high degree of thermal and chemical stability.
Tubes withstand centrifuged forces up to 3000 ×g and are suitable for sample transport or the culture of microorganisms.
Ordering information: Additional polyethylene stoppers are sold separately.
Delivery information: Tube 62409-465 is sterilized and supplied with a mounted stopper.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty