Would you like to log in again?
Nunc® 384-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nunc®
464718 264574 242765 264675 242764 265203 262260 265203 264573 264575 265202 265202 242765 264575 262160 4309 264573 242764 4315 264576 264579 460518 264579 242757 264675 242757 264576 460518 165195 464718 460372 460372 264574 262160 262260 165195 4308 4312
21993-902CS 1079.32 USD InStock
CA21993-912 21993-902 CA62409-602 CA21993-910 62409-634 82030-994 CA21993-914 82030-992 101504-474 21993-908 CA62409-064 CA62409-062 CA62409-060 CA73521-436 62409-060 CA73521-438 CA73521-434 21993-910 CA62409-604 21993-914 62409-604 CA62409-634 62409-602 CA82030-994 21993-912 CA82030-992 CA62409-454 CA21993-902 73520-634 CA21993-908 101504-460 101504-462 62409-064 62409-062 73521-438 101504-468 73521-434 73521-436
Nunc® 384-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
These high-density plates are designed for a variety of high throughput screening applications and drug discovery.
  • Clear plates have excellent optical clarity for read-through applications
  • White plates maximize reflection of signal and maintain low background for luminescence/fluorescence applications
  • Black plates reduce background and minimize backlight scatter for fluorescence assays
All plates have a pinchbar design
Plates are available untreated, for molecular biology applications, or with a MaxiSorp™ surface for immunology applications. MaxiSorp™ surface is recommended for assays where glycoproteins (i.e., antibodies) are absorbed into the plastic. Optical-bottom plates have a white or black upper structure with an optically clear polystyrene bottom. They can be read from the top or the bottom and are optimized for scintillation counting.
The polystyrene plates feature a flat bottom, while the polypropylene plates have a round bottom to facilitate sample retrieval. Plates feature rounded square wells to reduce well-to-well contamination, and have a total volume of 120µL per well. With the same footprint as 96-well plates, 384-well plates are compatible with robotics, automated readers, and liquid handling systems.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty