Would you like to log in again?
Nunc® Replication Systems, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
250393 250520 250393 250520
62409-608EA 4508.57 USD InStock
CA62409-608 62409-608 CA62409-606 62409-606
Nunc® Replication Systems, Thermo Scientific
Microplate and Petri Dish Replicators
Designed for use with 96-well and 384-well plates (see 62409-602 series) and OmniTrays (see 62409-600 series).
  • Pin replicators are ideal for transferring small volumes of inoculum from MicroWell™ plates to other plates or solid supports
  • Replicator pins are durable stainless steel and are precisely aligned to ensure long-term performance even after repeated flame sterilization
Biodyne® B nylon membranes (62409-614) are precut to fit into the OmniTray, for manual or automated arraying directly on a solid support.
MicroWell™ Plate Copier (62409-610) and OmniTray Copier (62409-612) precisely register the replicator pins to agar, hybridization membranes, or the wells of MicroWell plates. Copiers are machined from polypropylene and are resistant to solvents and bleach.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty