Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Palletote® Boxes, Rubbermaid®
Supplier: Rubbermaid Commercial Products
Rubbermaid®
RCP1721GRA RCP1731GRA RCP1731GRA
62661-363EA 108.65 USD InStock
62661-363 62661-125 62661-362
Palletote® Boxes, Rubbermaid®
Boxes Storage Boxes
Injection-molded high-density polyethylene provides excellent stress and crack resistance.
  • USDA tested
  • Ultra-durable
  • Slip-resistant textured bottom
  • Easy to clean
  • Nestable and stackable
  • Optional lid (sold separately)
Used for light duty in process operations and as returnable/reusable shipping containers. High stress crack resistance.
Certifications: USDA-tested storage box complies with FDA regulations regarding use in meat processing and food handling environments.
Ordering information: Boxes and lids sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty