Would you like to log in again?
AkroBins® Bin Dividers and Hanging Systems, Akro-Mils™
Supplier: Akro-Mils
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AkroBins®
30220YELLO 30220RED 30240BLUE 30210YELLO 30250YELLO 30-230R 30230BLUE 30240YELL 30210RED 30250RED 30-230Y 30210BLUE 30240RED 30250BLUE 30220BLUE 30235BLUE
62661-514CS 681.72 USD InStock
62661-514 62661-504 62661-516 62661-506 62661-518 62661-508 62661-520 62661-455 62661-532 62661-456 62661-522 62661-534 62661-512 62661-458 62661-524 62661-461
AkroBins® Bin Dividers and Hanging Systems, Akro-Mils™
Boxes Dispenser Boxes
Heavy-duty polypropylene AkroBins® bins are waterproof, rustproof, corrosion-resistant, and impervious to weak acids and alkalis.
  • Autoclavable
  • Designed with an anti-slide stop, reinforced ribs, and a wide ridge for multi-level stacking
Clear polystyrene lids keep products clean, dust-free, and visible. Bin dividers maximize space for storage of smaller parts and accessories. Bin panel 62661-490 and bench rack 62661-498 are specifically designed to organize AkroBins® bins. The 16-gauge steel louvered bin panels with gray baked enamel finish are mountable on almost any vertical surface. The freestanding bench rack with gray baked enamel finish provides heavy-duty desktop storage. Bin panels and bench racks can be used with any size AkroBins® bins.
Ordering information: Bins, lids, dividers, and supports are ordered separately.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty