Would you like to log in again?
ESD AkroBins®, Akro-Mils
Supplier: Akro-Mils
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AkroBins®
30210ESD 30240ESD 30220ESD 30235ESD 30230ESD
62661-550CS 2077.34 USD InStock
62661-550 62661-542 62661-536 62661-547 62661-538
ESD AkroBins®, Akro-Mils
Boxes ESD-Safe Boxes
Molded from CarmelStat™, these black ESD AkroBins® bins dissipate electrostatic charges for safe and secure storage of sensitive electronic components
  • Unaffected by weak acids and alkalis
  • Sturdy, heavy-duty, one-piece construction is water, rust, and corrosion-proof
  • Resistant to oil and grease
Containers feature a full-width hanger lip to support heavy loads when suspended from hanging rails or louvered panels. Reinforced side ribs provide added strength to prevent side-wall spreading when bins are stacked or loaded. Large label slot on front of bin provides easy bar code scanning and content identification. Anti-slide stop prevents bins from shifting forward when stacked.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty