Would you like to log in again?
CHEMWARE® Sterilization Tray, Saint Gobain Performance Plastics
Supplier: Saint-Gobain
Chemware®
D1069686
62670-051EA 465 USD InStock
62670-051
CHEMWARE® Sterilization Tray, Saint Gobain Performance Plastics
Trays/Dishes/Pots Sterilizing Trays/Pans
This tray can be used at high or cryogenic temperatures between –370 to 500 °F (–200 to 260 °C).
  • Chemical resistant
  • Excellent for corrosive and solvent baths
  • Low–and high–temperature resistant
Ideal for staining standard microscope slides or as a holding container when working with corrosive or solvent baths. Cover prevents evaporation of liquid contents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty