Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SP Bel-Art® Sampler Syringe, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37879-0000
62866-182CS 422.62 USD InStock
62866-182
SP Bel-Art® Sampler Syringe, Bel-Art Products, a part of SP
Samplers Fluid Samplers
This liquid sampler syringe is a polypropylene tube that can be placed into hot liquids up to 80 °C without fear of damage.
  • Excellent temperature resistance
  • Easy to use
  • Graduated markings
It features a graduated capacity of 30 cc (1 oz.) and squeeze bulb graduated to 100 mL in 5 mL increments.
Operation is simple: squeeze bulb to draw liquids into tube. If the liquid is not to be transferred into another vessel, the syringe can be inverted and the liquid will be contained in the graduated bulb.
Certifications: FDA approved materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty