Would you like to log in again?
SP Bel-Art Stepped Tubing Connectors, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
H19559-0000 H19558-0000 H19563-0000 H19562-0000 H19561-0000
16126-390PK 47.56 USD InStock
16126-390 16126-396 16126-388 16126-386 62866-401
SP Bel-Art Stepped Tubing Connectors, Bel-Art Products, a part of SP
Fittings and Connectors
These fittings ensure a tight, secure connection between tubing having different inner diameters.
  • Tight, secure connection
  • Polypropylene plastic construction
  • Chemically resistant
  • Autoclavable
Constructed of chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty