Would you like to log in again?
VWR® 99% Protein-Free Amber Tubing
Supplier: VWR
2008 404R
62996-695PK 454.46 USD InStock
62996-695 62996-380
VWR® 99% Protein-Free Amber Tubing
Tubing Transfer Tubing
Extensively leached, washed and surface treated to undetectable protein levels.
  • 99% protein-free
  • Amber color
Compatible with steam, ETO, or gamma radiation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty