Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® 50 Platinum-Cured Silicone Tubing for Peristaltic Pumps, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8060-3016 8060-3018 8060-3173 8060-3017 8060-3015 8060-3182 8060-3025 8060-3024
63015-318CS 292.72 USD InStock
63015-318 63015-314 63015-316 63015-328 63015-322 63015-324 63015-330 63015-320
Nalgene® 50 Platinum-Cured Silicone Tubing for Peristaltic Pumps, Thermo Scientific
Tubing Transfer Tubing
Flexible, durable, and translucent Platinum Silicone tubing is designed to provide extended pump life in peristaltic applications.

  • Tubing contains no phthalate plasticizers or peroxides that can leach out into transported fluid
  • Autoclavable

This high-purity tubing enables a variety of pump and transfer applications including pharmaceutical, laboratory, bioprocess manufacturing, and food and beverage.

Certifications: Tubing meets FDA, USDA, U.S. Pharmacopeia Class VI, 24th Edition; and 3A sanitary standards.

Ordering information: Supplied in 7.6m (25') coil lengths.

Caution: Note: Not intended for invasive use.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty