Would you like to log in again?
Nalgene® 890 Teflon® FEP Tubing, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
8050-0125 8050-0437 8050-0375 8050-0562 8050-0500 8050-0187 8050-0310 8050-0250
63014-726CS 848.33 USD InStock
63014-726 63014-718 63015-336 63014-722 63014-714 63015-332 63014-710 63015-334
Nalgene® 890 Teflon® FEP Tubing, Thermo Scientific
Tubing Transfer Tubing
Translucent, FEP, semi-rigid tubing for demanding lab applications such as chromatography, trace metal analysis, pollution sampling, highly reactive catalyst procedures, metallurgical corrosion testing, pharmaceutical work, dissolutions, and hot acid etchings.
  • Non cytotoxic and non-contaminating
Operating temperature range: –75 to +205 °C (–103 to +400 °F). Durometer hardness: Shore D, 58. Can be sterilized by autoclaving, gas, dry heat, and disinfectants. Do not radiation-sterilize. Resin complies with FDA Reg. 21 CFR 177.1550 for food contact.
Superior non-stick characteristics aid in the transferring of biological preparations where protein binding/adsorption is a problem. Also eases movement of viscous, aggressive compounds. For low-pressure use with mechanical, compression-type fittings.
Ordering information: Supplied in a 25' coil length or in a case of two coils.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty