Would you like to log in again?
SP Bel-Art Tubing Cutter, Plastic, Bel-Art Products, a part of SP
Supplier: Bel-Art
Scienceware®
H21010-0000
63022-100CS 501.52 USD InStock
63022-100
SP Bel-Art Tubing Cutter, Plastic, Bel-Art Products, a part of SP
Cutters
Pocket-sized cutter cuts flexible tubing with an outer diameter up to 19 mm (3/4").
  • Hardened steel blade
  • Jaw is spring-loaded and closes automatically
  • Compact and convenient size
The jaw is lightly spring-loaded and closes automatically for safety. The replaceable, hardened steel blade produces clean, straight cuts.
Ordering information: Replacement blades sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty