Would you like to log in again?
Coplin Staining Jar with Cover, Variety Glass
Supplier: Variety Glass
673 675
63025-010PK 430.04 USD InStock
63025-010 74280-016
Coplin Staining Jar with Cover, Variety Glass
Jars Staining Jars
Flint glass staining jar with ground-on cover
Holds ten 75x25mm (3x1") slides.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty