Would you like to log in again?
Precision Tweezers Series 800, Walter Stern
Supplier: Walter Stern
830-003 860-010
63039-990EA 87.48 USD InStock
63039-990 25607-101
Precision Tweezers Series 800, Walter Stern
Forceps
Stainless steel tweezers with very sharp points.
  • Anti-acid and antimagnetic
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty