This thick and medium pointed tweezer is excellent for general assembly.
- Strong tips, won't bend
- Most popular general assembly tweezer
The 'Style 00' Series straight medium pointed tweezers is the most popular general purpose tweezer used without magnification. The series features robust medium pointed tips for longevity and a variety of choices, like ergonomic handles, serrated tips, radius edges and fluted handles, depending on your application. Three different qualities are offered. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).