Would you like to log in again?
Excelta Corp® Roto-Pic™ Vacuum Pickup System
Supplier: Excelta
2000RK
63041-436EA 4878.09 USD InStock
63041-436
Excelta Corp® Roto-Pic™ Vacuum Pickup System
Vacuum Handling Systems
This system is Ideal for lifting, placing and turning components with the simple movement of one finger.
  • Index finger slide switch for on/off
  • Index finger wheel rotates tip 360°
  • Available with many tip size/configurations
The 2000RK five star vacuum system is the most revolutionary vacuum pick-up available. Designed to reduce manual handling of both chips and wafers, the Roto-Pic has a smooth positive hold-and-release slide bar. The round nose wheel rotates 360° so components can be picked up and aligned with ease.
Ordering information: Includes Roto-Pic 2000A, 2000B, 2000C, 2020, 2021, 2022, 2023, 2024, 2050, 2051, 2052, 2060, 2071, 2040, 2041, 2043, 2044, 2080. Spare components are available at accessories.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty