Would you like to log in again?
Tweezers, Broad Flat Wafer Handling, Excelta Corp®
Supplier: Excelta
35A-SA 36A-SA 25-SA 35A-SA-SE
63042-026EA 14.28 USD InStock
63042-026 63042-040 63042-024 63041-684
Tweezers, Broad Flat Wafer Handling, Excelta Corp®
Forceps
These are ideal for picking flat objects off a flat surface.
  • Large flat tip offers better control of item
  • Anti-magnetic stainless steel, cleanroom safe
  • Two different quality grades
These tweezers feature straight large flat round pointed tips and serrated handles for better control. They are made of anti-magnetic Neverust® stainless steel and is 4.75" long. We offer 2 different qualities. The higher the quality, the more precise the tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty