Would you like to log in again?
Spatulas, Miniature, Plastic Handle, Excelta Corp®
Supplier: Excelta
260K 260B EXA-260A 260D 260A 260D 260D-R 260B 260A-R 260A-R 260C 260C 260AA
76471-548EA 24.64 USD InStock
76471-548 75880-964 75880-878 76238-660 63041-692 63041-690 63041-696 63041-694 76240-492 75880-882 76250-658 76243-278 76238-658
Spatulas, Miniature, Plastic Handle, Excelta Corp®
Spatulas
These miniature spatulas are designed for placing epoxy/lubricants in very small amounts in a precise location.
  • Miniature design for accessibility under scope
  • Color coded width sizes for identification
  • Different tip widths to fit different applications
These mini-spatula (oiler) are 2.5" long with a plastic handle. Mini-spatulas are ideal for placing epoxies or lubricant in very small amounts into tight places. They feature different size tips (color coated for identification).
Spatula shape: Flat end blade/straight handle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty