Would you like to log in again?
Brushes, Scratch, Replaceable Tips, Excelta Corp®
Supplier: Excelta
265 266 268 264 263 267
63041-742EA 22.54 USD InStock
63041-742 63041-710 63041-738 63041-714 63041-730 75880-970
Brushes, Scratch, Replaceable Tips, Excelta Corp®
Brushes
Used for cleaning and polishing, these dial-out bristles can be stiff or soft, depending on how far they protrude from the handle.
  • Replaceable bristles
  • Bristles 'dial-out' for different stiffness
  • Four different bristle materials
These scratch brushes feature replaceable bristles with a plastic handle. We offer bristles of brass, steel, fiberglass and nylon. Simple twist the collar and the bristles appears. It can be dialed out, to your preference, based on how soft or hard you want the bristles.
Delivery information: The kit includes one handle and 1 each brass, steel, fiberglass and nylon refill in a plastic case.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty