Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Probes, Fine Tip, Stainless Steel, Excelta Corp®
Supplier: Excelta
333F 331D 330A 331B 331C 334 330D 334D 331D 334C 330C 330K 331A 330B
63041-942EA 10.16 USD InStock
63041-942 63041-932 75880-588 63041-940 63041-946 75880-920 63041-958 63041-936 63041-944 63041-956 63041-934 63041-950 76240-220 63041-938
Probes, Fine Tip, Stainless Steel, Excelta Corp®
Dissection Needles
These 0.01" probes are ideal for fine work.
  • Cleanroom safe, stainless steel
  • 3" overall length for work under a scope
This series three star fine straight tip stainless steel probe has 10 mil diameter tip for precise work. They are offered with a standard 6" overall length and also in a 3" version.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty