Would you like to log in again?
Mixing Spatula, Excelta Corp®
Supplier: Excelta
345
63041-982EA 12.62 USD InStock
63041-982
Mixing Spatula, Excelta Corp®
Spatulas
The 345 two star stainless steel 8" long spatula with wooden handle can be used for mixing and potting.
  • Spatula shape: Rounded end blade/straight handle
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty