Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tweezers, 4" Wafer Handling, Excelta Corp®

Supplier: Excelta
491D-SA-PI 490B-SA-PI 491P-SA-PI 490-SA-PI 490T-SA-PI 491-SA-PI 490L-SA-PI
63042-356EA 115.7 USD InStock
63042-356 63042-346 63042-358 63042-348 63042-350 63042-354 63042-344
Tweezers, 4" Wafer Handling, Excelta Corp®
Forceps
These tweezers are specifically designed handling 4" wafers.

  • For scratch free holding of 4" wafers
  • Cleanroom safe, anti-mag. Stainless steel
  • Various configurations for various applications

The wafer tweezers for 4" wafers are made of stainless steel and are anti-magnetic. They come in a variety of configurations. The 'B' version features a 20° bend in the body to improve handling. The overall length is 4.75".

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty