Would you like to log in again?
Tweezers, Straight Taper Relieved Tips, Style 4, Excelta Corp®
Supplier: Excelta
4-SA-PI 4-SA-ET 4-SA-SE 4-SA 4-SA
76238-694EA 124.58 USD InStock
76238-694 63042-368 89411-846 63042-372 63042-374
Tweezers, Straight Taper Relieved Tips, Style 4, Excelta Corp®
Forceps
Designed for microscopy, the tapered tips offer excellent visibility under a scope.
  • Specifically made for microscopy
  • Available in three different materials
  • Three different quality grades available
  • Unsurpassed visibility under a scope
  • Three quality grades available
The 'Style 4' straight tapered ultra-fine pointed tweezers is primarily used for microscopy. The overall length is 4.25". These are also availabe with ergonomic handles (ET). We offer 3 different qualities. The higher the quality, the more precise the tips. They are made of stainless anti-magnetic steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty