Would you like to log in again?
Pliers, Wide Nose, Excelta
Supplier: Excelta
508C-US 508-US 508D-US 508B-US
75880-924EA 453.84 USD InStock
75880-924 63042-492 63042-494 63042-496
Pliers, Wide Nose, Excelta
Pliers
The 508 series will straightens multiple lead wires in one easy operation.
  • Wide nose ideal for large items
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These wide nose straightening pliers uniquely straighten multiple leads in one easy operation. The jaw inserts are factory replaceable and available in carbon steel, brass, or acetal. The overall length is 5.5". It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The shank is made of carbon steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty