Would you like to log in again?
Pliers, Replaceable Tip, Straight, Excelta
Supplier: Excelta
532B-US 533-US 531-US 532-US 2833 531A-US 531B-US 2831
63042-554EA 316.45 USD InStock
63042-554 63042-556 63042-558 75880-934 63042-560 63042-552 89411-774 63042-564
Pliers, Replaceable Tip, Straight, Excelta
Pliers
These pliers are useful in straightening leads and wires when metal-to-metal contact is not desired.
  • Anti-shock trimming of leads to specific length
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
This series of pliers offer soft jaw inserts to protect delicate parts. The jaw inserts are factory replaceable, and offered in Acetal, Brass and Nylon. The overall length is 5.75". They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty