Would you like to log in again?
Three Star™ Tweezers, Anti-Crush Devices, Excelta Corp®
Supplier: Excelta
53C-SA 52A-SA
63042-518EA 96.29 USD InStock
63042-518 63042-570
Three Star™ Tweezers, Anti-Crush Devices, Excelta Corp®
Forceps
Outfitted with an 'anti-crush' devise, these tweezers define the amount of pressure put on a delicate component.
  • Anti-crush device protects delicate items
  • Cleanroom safe
  • Made of anti-magnetic stainless steel
The tweezer tips collapse when excessive force is placed on the tweezer.
These two tweezers are designed for protecting delicate items. The anti-crush device allows the tweezer tips to collapse when too much pressure is applied. They are made of anti-magnetic Neverust® stainless steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty