Would you like to log in again?
Tweezers, Straight Taper Relieved Tips, Style 5, Excelta Corp®
Supplier: Excelta
5-SA-SE 5-S 5-SA-ET 5-SA-ET 5-S-ET 5-S-PI 5-SA MW5-SA 5-SA-PI-ET 5-S-SE 5-SA-PI 5 5-SA-PI-ET 5-SA
10066-482EA 110.93 USD InStock
10066-482 63042-380 63042-698 63042-688 63042-700 89411-782 63042-702 89411-854 63042-690 76238-704 63042-692 76238-698 76238-700 63042-696
Tweezers, Straight Taper Relieved Tips, Style 5, Excelta Corp®
Forceps
The style 5 tweezers are tapered and relieved at the tip to make them ideal for work under high magnification.
  • Specifically made for microscopy.
  • Available in three different materials
  • Three different quality grades available
  • Unsurpassed visibility under a scope
  • Three quality grades available
The '5-SA Style' series tweezers features straight tapered ultra-fine points and is primarily used for microscopy. The style 5 has the finest points available. The overall length is 4.25". It is made of anti-magnetic Neverust® stainless steel. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). The MW5-SA has radius inside edges to protect from scratching delicate items.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty