Would you like to log in again?
Tweezers, Long Very Fine Tip, Excelta Corp®
Supplier: Excelta
60-SA SS-S SS-SA SS-SA-ET 65A-SA SS-SA-SE SS-S-SE
63042-720EA 100.67 USD InStock
63042-720 89411-694 63042-712 63043-690 89411-814 89411-696 89411-830
Tweezers, Long Very Fine Tip, Excelta Corp®
Forceps
These tweezers are ideal for working in confined areas where a 'long reach' and precision is necessary.
  • Very fine points for precise work
  • Extra long handle for hard to reach places
  • Available in several different quality grades
These long tweezer feature very fine tips and great accessibility in tight places. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel. The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty