Would you like to log in again?
Cutters, Angulated Head, Excelta
Supplier: Excelta
7175E 7282E 7204E 7182E 7376E 90EI 73E-I 7104E 7375E 7172E+ 7275E 67EI 7272E
75880-696EA 149.63 USD InStock
75880-696 76471-524 76471-628 63042-816 76471-630 76471-520 76471-522 94024-320 63042-876 63042-810 63042-868 63042-814 63042-872
Cutters, Angulated Head, Excelta
Cutters
The angled head offers excellent visibility and accessibility to small wires and leads.
  • Angled cutting blades for difficult areas
  • 'Set-screw' stop prevents overtravel
  • Induction hardened blades for longevity
  • Ergonomic handles and dual leaf springs
The Lazer Line® angulated/tapered maximum flush cutters offer excellent visibility/accessibility in difficult areas. They are all made of carbon tool steel with induction hardened cutting blades to RHC 63-65. These tools feature angled tips with a set-screw to prevent over-travel while prolonging the life of the tool. The grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq. The dual leaf springs provide smooth action.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty