Would you like to log in again?
Five-Star™ Cutters, Medium Head, Excelta
Supplier: Excelta Corporation
Five Star™
7247E 7146E 7242E 7240E 7148E 7241E 7140E 7244E 7245E 7142E 7246E 7144E 7147E 7143E 7248E 7141E 7145E+
63042-848EA 144.23 USD InStock
63042-848 63042-838 63042-796 63042-786 63042-798 63042-842 63042-788 63042-844 63042-834 63042-846 63042-836 63042-790 63042-792 63042-782 63042-794 63042-850 63042-784
Five-Star™ Cutters, Medium Head, Excelta
Cutters
Five-Star™ 7000 series medium head cutters are precision side cutters designed to make an extremely precise cut on small leads and soft wire.
  • Available either with long foam cushion grips or ESD safe molded grips
  • The dual stainless-steel leaf springs provide smooth action
These are feature with raised bearing surface, Rockwell C 63-65 hardness, and a Delrin washer for smooth operation. The CNC machining allows the joint surfaces to be perfect to allow for long life.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty