Would you like to log in again?
Tweezers, Straight Fine Tips, Boley Style, Excelta Corp®
Supplier: Excelta
MM-SA-SE MM AA-SA-PI AA-SE AA-SA AA-SA-PI AA-SA-SE MM-SE AA-SA-PI-ET AA AA-S AA-SA-SE-PH AA-SA-ET MM-SA-PI GG-SA AA-S-SE
63043-210EA 12.27 USD InStock
63043-210 63043-484 63043-198 63043-482 63043-480 63043-208 76239-950 63043-214 63043-204 63043-212 63043-202 89411-710 89411-822 89411-866 89411-768 89411-752
Tweezers, Straight Fine Tips, Boley Style, Excelta Corp®
Forceps
Designed for general assembly.
  • Strong tips for general assembly use
  • Available in different materials for different applications
  • Three different quality grades available
  • Option of ergonomic grips
These 'BOLEY' straight medium point general purpose tweezer are offered in 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat. The 'PH' suffix denotes plastic coated handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty