Would you like to log in again?
Four Star™ Tweezer, Titanium, EXCELTA®
Supplier: Excelta
7-TA 5-TA
63043-380EA 191.58 USD InStock
63043-380 63043-698
Four Star™ Tweezer, Titanium, EXCELTA®
Forceps
These tweezers are made from lightweight titanium alloy and are great for all solder applications (solder will not adhere to the tip)
They are heat resistant to 806°F.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty