Would you like to log in again?
North™ Butyl Gloves
Supplier: North by Honeywell
North™
B074GI/S B074GI/S B074GI/XS B074GI/XS B074GI/L B074GI/L B074GI/M B074GI/M
63550-053CS 9460 USD InStock
63550-053 63550-055 77532-368 77532-370 63550-057 63550-059 77532-374 77532-372
North™ Butyl Gloves
Gloves
These black butyl gloves are chemical resistant.
  • Flexible and sensitive, even at lower temperatures
  • Rolled bead cuffs with smooth finish
  • Unlined with sanitized interior
Butyl has the highest permeation resistance to gas and water vapor, highly resistant to ketones (MEK, MIBL, acetone) and esters (tricresyl phosphate, amyl acetate, ethyl acetate).
Work applications include aircraft maintenance, chemical handling and processing, domestic preparedness, laboratory and HazMat.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty