Would you like to log in again?
MILSPEC-Type Butyl Gloves, Honeywell Safety
Supplier: Honeywell Safety
B254GI/XL B254GI/L B144RGI/XS B144RGI/L B144GI/M B144GI/XS B254GI/M B144RGI/XL B144GI/XL B144RGI/M B144RGI/S B144GI/S B144GI/L
63550-081PR 123.27 USD InStock
63550-081 63550-071 63550-073 63550-063 63550-097 63550-075 63550-065 63550-099 63550-077 63550-067 63550-101 63550-079 63550-069
MILSPEC-Type Butyl Gloves, Honeywell Safety
Gloves
Gloves are highly impermeable to water vapor, gases, and toxic chemicals including most missile fuels, and resistant to oxygenated solvents and most oxidizing chemicals.
  • Curved hand design
  • Gloves 63550-102, -104, -106, -108, and -110 feature a cotton liner
They are flexible and sensitive, even at low temperatures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty