Would you like to log in again?
Dewar Vacuum Bottles, Shallow-Form, Pope Scientific
Supplier: Pope Scientific
8335 8345 8130 8315 8325 8110 8120
63884-971EA 657.59 USD InStock
63884-971 63884-973 63884-961 63884-975 63884-963 63884-965 63884-969
Dewar Vacuum Bottles, Shallow-Form, Pope Scientific
Dewars and Cryogenic Vessels
Shallow-form Dewars permit cooling operations not practical with standard cylindrical or spherical Dewars
Available with hemispherical or flat bottom interiors in protective aluminum base.
These vacuum bottles are made of borosilicate glass.
Applications include rapid freezing and recovery of tissue/organ specimens, insertion of small flasks, agitation in flask with magnetic stirrer, mixing and tamping with stirring rods or pestles. Cooling range is from dry ice/acetone to liquid nitrogen (LN2). Manufactured for use with liquid nitrogen and ideal for use with liquid gases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty