Would you like to log in again?
Valve Port Hat-Type Specialty Needles, Hamilton Company
Supplier: Hamilton
90825 90815 90816 90826 90821 90824 90822 90818 90817 90823 90820 90819
89221-526PK 75.59 USD InStock
89221-526 89221-538 89221-528 89221-544 89221-534 89221-536 89221-540 89221-530 89221-542 89221-532 65515-061 65515-141
Valve Port Hat-Type Specialty Needles, Hamilton Company
Syringe Needles
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry
Hat-type needles are designed to be connected directly to a flat bottom valve port. Use the Hamilton Needle Bushing part number 35056 to connect the needle directly to any 1/4-28 Hamilton valve. Custom Length and Gauge
Each specialty needle is made to order. Custom lengths range from 0.375 to 12 inches. Available point styles include 2, 3 and 4. Please specify the desired length and point style when ordering.
Needle Point Styles (Custom lengths and point styles available)
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10.
Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10.
Point Style 4 - Point Style 4 - Standard 12° beveled needle point is recommended for life science applications. Available gauges: 34-10. Special point styles such as 30°, 45°, or any other angle are available upon request.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Ordering information: If you are looking for a needle point, gauge or length that is available but not listed here, please call Avantor to place a special order by phone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty