Would you like to log in again?
SP Bel-Art Beta-Safe™ Storage Box, Bel-Art Products, a part of SP
Supplier: Bel-Art
SP Bel-Art Beta-Safe™
F24987-0000
66001-132EA 568.73 USD InStock
66001-132
SP Bel-Art Beta-Safe™ Storage Box, Bel-Art Products, a part of SP
Boxes Beta/Gamma Radiation Boxes
Fabricated from 9.5mm (⅜") clear acrylic for all around visibility of stored contents, this box stores radioactive materials such as P-32, S-35 and C-14
  • Hinged lid with hasp, may be secured by a locking ¼" cable through anti-theft connectors that are securely attached to the box
The standard unit measures a generous 25.4 x 15.24 x 17.8cmH (10 x 6 x 7H") I.D.
A hinged lid with a hasp is provided for locking. The entire unit may be secured by utilizing a locking ¼" cable through the anti-theft connectors which are securely attached to the box.
Certifications: Complies with USNRC Rules and Regulations, Title 10, Chapter 1, Part 20.
Ordering information: Other sizes available on special order. Lock and cable or chain not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty