Would you like to log in again?
Straight Sided Stoppers, DWK Life Sciences
Supplier: DWK Life Sciences
73811-21 73811T-13 73811T-21
66004-000CS 150.73 USD InStock
66004-000 66004-006 66004-004
Straight Sided Stoppers, DWK Life Sciences
Stoppers Rubber Stoppers
These high quality gray chlorobutyl stoppers are used for research and pharmaceutical packaging applications.
  • Formulation contains no plasticizers, 2-mercapto-benzothaizole, nitrosamine precursors or natural rubber latex
  • Formulation is applicable to aqueous solutions with a pH range of 2 to 10
  • Durometer 50
  • Universal gray chlorobutyl formulation passes Japanese, European and United States pharmacopeia testing for globally marketed pharmaceutical products
*Is rinsed with a silicon oil to provide increased lubricity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty