Would you like to log in again?
Aluminum Seals, Tear-Out, Kimble Chase
Supplier: DWK Life Sciences
73820-20 73820B-13 73820B-20 73820C-20 73820D-13 73820C-13 73820D-20 73820-13
66004-030CS 198.92 USD InStock
66004-030 66004-032 66010-795 66010-797 66004-028 66004-034 66004-024 66004-026
Aluminum Seals, Tear-Out, Kimble Chase
Closures Chromatography Vial Closures
These tear-out seals are designed to secure septa and stoppers to bottles and vials.
  • Tamper-evident center tear-out feature protects target area
  • For use with 13 and 20 mm serum finished bottles and vials
  • Allow for easy access to septa
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty