Would you like to log in again?
CASE EQUIPMENT PROTECTOR BLK
Supplier: PELICAN PRODUCTS
Pelican* Protector Case*^ Unbreakable, watertight, dustproof, chemical-resistant, and corrosion-proof. Made of ultra-high impact structural copolymer for durability. 6.4mm neoprene O-ring and ABS latches. Temperature: -23 to 99[degree]C. Outside: 40.6x33x17.4cm. Depth: 10.8cm. Black.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CASE EQUIPMENT PROTECTOR BLK 66005-660 1450-000-110 Each Retrieving
Pack of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty