Would you like to log in again?
VIAL KITS AMBR W/CAP+SEP PK100
Supplier: Thermo Scientific
Assembled 8-425 Glass Vial Kits^ Vials, caps and septa are assembled, reducing possible contamination of the vial. Vials have a 1.5 ml nominal volume. Vial: Amber Glass. Closure: Black Polypropylene. Septum: Red PTFE/Silicone, 0.060"
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL KITS AMBR W/CAP+SEP PK100 66008-062 200 406 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty