Would you like to log in again?
Nalgene® Bulk-Packed, Non-Sterile, Cryogenic Vials, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5011-0020 5011-0012
66008-748CS 669.67 USD InStock
66008-748 66008-745
Nalgene® Bulk-Packed, Non-Sterile, Cryogenic Vials, Thermo Scientific
Vials
Nalgene cryogenic vials are externally threaded vial for aseptic technique, using a design similiar to the linerless closures used on Nalgene bottles and carboys.
  • Self-standing and may be used without a holder
They are an economical, non-sterile version of Cat. Nos. 5000-0012 and -0020 without printing or graduations.
Ordering information: Closures and vials are packed separately.
Caution: For research use only; not for in-vitro diagnosis or parenterals. Do not autoclave.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty