Would you like to log in again?
Prescored Ampules, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
176796 176780 176779 176776 176792 176782 176772 176799
66009-020CS 98.31 USD InStock
66009-020 66009-040 66009-024 66009-022 66009-028 66009-006 66009-061 66009-083
Prescored Ampules, Wheaton®, DWK Life Sciences
Ampules
Clear or amber prescored ampules: Clear ampules are manufactured from Wheaton 33® brand borosilicate glass. To protect light-sensitive samples, amber ampules are manufactured from Wheaton 320 brand amber borosilicate glass.
  • Wheaton prescored ampules eliminate filing
  • Stems can be pull-sealed or tip-sealed to create an effective hermetically sealed container
  • A Gold Band® identifies the prescored feature
Available in different sizes.
Certifications: Clear: USP Type I standards; Amber: USP Type I and ASTM Type I, Class B standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty