Would you like to log in again?
White Urea Closures with PTFE-Faced White Rubber Liners, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
74526-22400
66009-258CS 630.35 USD InStock
66009-258
White Urea Closures with PTFE-Faced White Rubber Liners, Kimble Chase, DWK Life Sciences
Closures Bottle Closures
Closure is made from white urea and features a foam-backed F-217 PTFE liner which resists attack from virtually all chemicals at room temperature.
  • Top is suitable for marking
  • Ideal for scintillation vials
For long term sample storage with resilience for a tight seal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty