Would you like to log in again?
KIMAX® Sample Vials, Borosilicate Glass, with Rubber-Lined Screw Cap, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
60940D-1 60940D-2 60812D-12 60940D-4 60940D-12 60940D-6 60940D-3
66010-423CS 1740.61 USD InStock
66010-423 89201-818 66010-425 66010-427 66010-426 66010-429 66010-428
KIMAX® Sample Vials, Borosilicate Glass, with Rubber-Lined Screw Cap, Kimble Chase, DWK Life Sciences
Vials
Sample vials with rubber-lined solid top screw caps unattached made of borsilicate glass.
  • Clear screw thread sample vials
  • Closed-top black phenolic closures with rubber liners, unattached
  • Economical and time-saving in the lab
Certifications: Low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty