Would you like to log in again?
KIMBLE® ACCUFORM® Graduated High Recovery Vials with Black Closures and PTFE-Faced White Rubber Liners, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
60702-5 60707-5 60702-2 60707-3 60702-1 60702-3 60707-1 60702-10 60707-2
66010-494CS 295.02 USD InStock
66010-494 66010-493 66010-496 66010-495 66010-498 66010-497 66010-490 66010-492 66010-491
KIMBLE® ACCUFORM® Graduated High Recovery Vials with Black Closures and PTFE-Faced White Rubber Liners, DWK Life Sciences
Vials
These vials are graduated to ensure more accurate measurements and dispensing.
  • Tapered bases allow for maximum sample recovery
  • Solid-top closure and PTFE-faced, white rubber liner, assembled and attached
Clear vials manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Amber vials manufactured from 51 expansion borosilicate amber glass conforming to USP, Type I requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty